Ethyl-2-cyano-2-(pyrimidin-2-yl)acetate 95%

Supplier: Apollo Scientific

Synonyms: Ethyl 2-cyano-2-(pyrimidin-2-yl)acetate, Cyanopyrimidin-2-yl-acetic acid ethyl ester

OR904774-1G OR904774-250MG
APOSOR904774-1GEA 137 EUR
APOSOR904774-1G APOSOR904774-250MG
Ethyl-2-cyano-2-(pyrimidin-2-yl)acetate 95%
Ethyl-2-cyano-2-(pyrimidin-2-yl)acetate
Formula: C₉H₉N₃O₂
MW: 191,1900 g/mol
Storage Temperature: Ambient
MDL Number: MFCD00297325
CAS Number: 65364-63-8

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR