Methyl quinoxaline-5-carboxylate 95%
Supplier: Apollo Scientific
OR904780-1G
OR904780-250MG
APOSOR904780-1GEA
105
EUR
APOSOR904780-1G
APOSOR904780-250MG
Methyl quinoxaline-5-carboxylate 95%
Methyl quinoxaline-5-carboxylate
Formula:
C₁₀H₈N₂O₂ MW: 188,1800 g/mol Storage Temperature: Ambient |
MDL Number:
MFCD19703859 CAS Number: 6924-71-6 |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...