Methyl-5-(tert-butyl)isoxazole-3-carboxylate 95%

Supplier: Apollo Scientific

Synonyms: Methyl 5-(tert-butyl)isoxazole-3-carboxylate

OR904812-1G OR904812-250MG
APOSOR904812-1GEA 734 EUR
APOSOR904812-1G APOSOR904812-250MG
Methyl-5-(tert-butyl)isoxazole-3-carboxylate 95%
Methyl-5-(tert-butyl)isoxazole-3-carboxylate
Formula: C₉H₁₃NO₃
MW: 183,2000 g/mol
Storage Temperature: Ambient
MDL Number: MFCD03778975
CAS Number: 517870-22-3

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR