6-Chloro-3-methylpyrimidin-4(3H)-one 98%

Supplier: Apollo Scientific

OR905192-1G OR905192-250MG
APOSOR905192-1GEA 807 EUR
APOSOR905192-1G APOSOR905192-250MG
6-Chloro-3-methylpyrimidin-4(3H)-one 98%
6-Chloro-3-methylpyrimidin-4(3H)-one
Formula: C₅H₅ClN₂O
MW: 144,5600 g/mol
Storage Temperature: Ambient
MDL Number: MFCD00955650
CAS Number: 101079-62-3

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR