7-Methyl-1H-indazole-3-carbaldehyde 95%
Supplier: Apollo Scientific
OR905208-1G
OR905208-250MG
APOSOR905208-1GEA
350
EUR
APOSOR905208-1G
APOSOR905208-250MG
7-Methyl-1H-indazole-3-carbaldehyde 95%
7-Methyl-1H-indazole-3-carbaldehyde
Formula:
C₉H₈N₂O Storage Temperature: Ambient |
MDL Number:
MFCD09027036 CAS Number: 1000340-51-1 |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...