5-((Trimethylsilyl)ethynyl)pyrimidine 95%

Supplier: Apollo Scientific

OR905251-1G OR905251-250MG
APOSOR905251-1GEA 178 EUR
APOSOR905251-1G APOSOR905251-250MG
5-((Trimethylsilyl)ethynyl)pyrimidine 95%
5-((Trimethylsilyl)ethynyl)pyrimidine
Formula: C₉H₁₂N₂Si
Storage Temperature: Ambient
MDL Number: MFCD09258881
CAS Number: 216309-28-3

Order Now

SPECIFICATIONS

Environmentally Preferable

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR