5-Methyl-1,2,3,4-tetrahydroquinoline hydrochloride 95%
Supplier: Apollo Scientific
OR905389-1G
OR905389-250MG
APOSOR905389-1GEA
163
EUR
APOSOR905389-1G
APOSOR905389-250MG
5-Methyl-1,2,3,4-tetrahydroquinoline hydrochloride 95%
5-Methyl-1,2,3,4-tetrahydroquinoline hydrochloride
Formula:
C10H14ClN MW: 183,6800 g/mol Storage Temperature: Ambient |
MDL Number:
MFCD09026775 CAS Number: 861368-85-6 |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...