Ethyl-3-amino-3-methylbutanoate hydrochloride 95%
Supplier: Apollo Scientific
Synonyms:
Ethyl 3-amino-3-methylbutanoate hydrochloride
OR905460-10G
OR905460-25G
APOSOR905460-10GEA
101
EUR
APOSOR905460-10G
APOSOR905460-25G
Ethyl-3-amino-3-methylbutanoate hydrochloride 95%
Ethyl-3-amino-3-methylbutanoate hydrochloride
Formula:
C₇H₁₆ClNO₂ MW: 181,6604 g/mol Melting Pt: 101 °C Storage Temperature: Ambient |
MDL Number:
MFCD11844787 CAS Number: 85532-40-7 |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...