1-Ethyl-2-pyrrolidone 96%
Supplier: Apollo Scientific
Synonyms:
1-Ethyl-2-pyrrolidinone, N-Ethyl-2-pyrrolidone, 1-Ethylpyrrolidin-2-one
OR905748-100G
OR905748-1KG
OR905748-500G
APOSOR905748-100GEA
53.9
EUR
APOSOR905748-100G
APOSOR905748-1KG
APOSOR905748-500G
1-Ethyl-2-pyrrolidone 96%
1-Ethyl-2-pyrrolidone
Formula:
C₆H₁₁NO MW: 113,16 g/mol Storage Temperature: Ambient |
MDL Number:
MFCD00003199 CAS Number: 2687-91-4 |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...