3-Benzylphenol 95%

Supplier: Apollo Scientific

OR905784-1G OR905784-250MG
APOSOR905784-1GEA 132 EUR
APOSOR905784-1G APOSOR905784-250MG
3-Benzylphenol 95%
3-Benzylphenol
Formula: C13H12O
Storage Temperature: Ambient
MDL Number: MFCD00036116
CAS Number: 22272-48-6

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR