Ethyl-2-(1-hydroxycyclohexyl)acetate 95%
Supplier: Apollo Scientific
Synonyms:
Ethyl 2-(1-hydroxycyclohexyl)acetate
OR905800-1G
OR905800-250MG
APOSOR905800-1GEA
62.5
EUR
APOSOR905800-1G
APOSOR905800-250MG
Ethyl-2-(1-hydroxycyclohexyl)acetate 95%
Ethyl-2-(1-hydroxycyclohexyl)acetate
Formula:
C₁₀H₁₈O₃ MW: 186.2500 g/mol Storage Temperature: Ambient |
MDL Number:
MFCD00095227 CAS Number: 5326-50-1 |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...