2,3-Dimethoxyquinoxaline 95%

Supplier: Apollo Scientific

OR905834-1G OR905834-250MG
APOSOR905834-1GEA 683 EUR
APOSOR905834-1G APOSOR905834-250MG
2,3-Dimethoxyquinoxaline 95%
2,3-Dimethoxyquinoxaline
Formula: C₁₀H₁₀N₂O₂
MW: 190,2 g/mol
Storage Temperature: Ambient
MDL Number: MFCD00068701
CAS Number: 6333-43-3

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR