3-Bromo-6-methylpyridazine

Supplier: Apollo Scientific

OR905842-25G
APOSOR905842-25GEA 482 EUR
APOSOR905842-25G
3-Bromo-6-methylpyridazine
3-Bromo-6-methylpyridazine
Formula: C5H5BrN2
MW: 173,0106 g/mol
Storage Temperature: Ambient
MDL Number: MFCD09831907
CAS Number: 65202-58-6

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR