Benzyl-4-hydroxypiperidine-1-carboxylate 97%

Supplier: Apollo Scientific

Synonyms: 1-Benzyloxycarbonyl-4-hydroxypiperidine, 4-Hydroxy-1-piperidinecarboxylic acid Benzyl ester, 1-Cbz-4-hydroxypiperidine, 1-Carbobenzoxy-4-piperidinol, 1-Carbobenzoxy-4-hydroxypiperidine, Benzyl 4-Hydroxy-1-piperidinecarboxylate, 1-Benzyloxycarbonyl-4-piperidinol, 1-Cbz-4-piperidinol

OR9059-100G
APOSOR9059-100GEA 58 EUR
APOSOR9059-100G
Benzyl-4-hydroxypiperidine-1-carboxylate 97%
Benzyl-4-hydroxypiperidine-1-carboxylate
Formula: C₁₃H₁₇NO₃
MW: 235,29 g/mol
Boiling Pt: 167 °C/0,2mmHg
Density: 1,554 g/cm³ (25 °C)
Flash Pt: >110 °C
Storage Temperature: Ambient
MDL Number: MFCD01863722
CAS Number: 95798-23-5

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR