1-Boc-4-cyanopiperidine 98%
Supplier: Apollo Scientific
|
Warning
|
Synonyms:
tert-Butyl 4-cyanopiperidine-1-carboxylate, N-BOC-Piperidine-4-carbonitrile
OR9060-5G
OR9060-25G
APOSOR9060-5GEA
21.9
EUR
APOSOR9060-5G
APOSOR9060-25G
1-Boc-4-cyanopiperidine 98%
1-Boc-4-cyanopiperidine
Formula:
C₁₁H₁₈N₂O₂ MW: 210.27 g/mol Melting Pt: 60…63 °C Storage Temperature: Refrigerator |
MDL Number:
MFCD01861223 CAS Number: 91419-52-2 UN: 3077 ADR: 9+EHS III |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...