6-Chloro-1H-pyrazolo[3,4-d]pyrimidine 96%

Supplier: Apollo Scientific

OR906012-1G OR906012-250MG
APOSOR906012-1GEA 155 EUR
APOSOR906012-1G APOSOR906012-250MG
6-Chloro-1H-pyrazolo[3,4-d]pyrimidine 96%
6-Chloro-1H-pyrazolo[3,4-d]pyrimidine
Formula: C₅H₃ClN₄
MW: 154,5571 g/mol
Storage Temperature: Ambient
MDL Number: MFCD10697157
CAS Number: 23002-51-9

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR