5-Methoxyisobenzofuran-1,3-dione 95%

Supplier: Apollo Scientific

OR906027-1G OR906027-250MG
APOSOR906027-1GEA 78.6 EUR
APOSOR906027-1G APOSOR906027-250MG
5-Methoxyisobenzofuran-1,3-dione 95%
5-Methoxyisobenzofuran-1,3-dione
Formula: C9H6O4
MW: 178,1415 g/mol
Storage Temperature: Ambient
MDL Number: MFCD02670942
CAS Number: 28281-76-7

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR