4-Amino-5-bromopyrimidine 97%

Supplier: Apollo Scientific

OR906160-1G OR906160-5G
APOSOR906160-1GEA 85.8 EUR
APOSOR906160-1G APOSOR906160-5G
4-Amino-5-bromopyrimidine 97%
4-Amino-5-bromopyrimidine
Formula: C₄H₄BrN₃
MW: 174,00 g/mol
Storage Temperature: Ambient
MDL Number: MFCD00233962
CAS Number: 1439-10-7

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR