Bicyclo[4.2.0]octa-1,3,5-trien-7-one 95%
Supplier: Apollo Scientific
OR906275-1G
OR906275-250MG
APOSOR906275-1GEA
344
EUR
APOSOR906275-1G
APOSOR906275-250MG
Bicyclo[4.2.0]octa-1,3,5-trien-7-one 95%
Bicyclo[4.2.0]octa-1,3,5-trien-7-one
Formula:
C₈H₆O Storage Temperature: Ambient |
MDL Number:
MFCD01652340 CAS Number: 3469-06-5 |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...