2-Amino-5-ethylpyrazine 95%
Supplier: Apollo Scientific
OR906857-1G
OR906857-250MG
APOSOR906857-1GEA
819
EUR
APOSOR906857-1G
APOSOR906857-250MG
2-Amino-5-ethylpyrazine 95%
2-Amino-5-ethylpyrazine
Formula:
C₆H₉N₃ Storage Temperature: Ambient |
CAS Number:
13535-07-4 |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...