2-Amino-4-piperidino-6-methylpyrimidine 95%
Supplier: Apollo Scientific
OR907039-10G
OR907039-1G
OR907039-5G
APOSOR907039-10GEA
806
EUR
APOSOR907039-10G
APOSOR907039-1G
APOSOR907039-5G
2-Amino-4-piperidino-6-methylpyrimidine 95%
2-Amino-4-piperidino-6-methylpyrimidine
Formula:
C₁₀H₁₆N₄ MW: 192.26 g/mol Storage Temperature: Ambient |
MDL Number:
MFCD01011781 CAS Number: 91717-22-5 |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...