6-Ethoxy-1H-indole 95%

Supplier: Apollo Scientific

OR907048-100MG OR907048-1G OR907048-250MG
APOSOR907048-100MGEA 48 EUR
APOSOR907048-100MG APOSOR907048-1G APOSOR907048-250MG
6-Ethoxy-1H-indole 95%
6-Ethoxy-1H-indole
Formula: C₁₀H₁₁NO
MW: 161.2000 g/mol
Storage Temperature: Ambient
MDL Number: MFCD00051498
CAS Number: 37865-86-4

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR