1-Pyridin-4-ylpiperidin-4-one 95%
Supplier: Apollo Scientific
OR907054-1G
OR907054-5G
APOSOR907054-1GEA
116
EUR
APOSOR907054-1G
APOSOR907054-5G
1-Pyridin-4-ylpiperidin-4-one 95%
1-Pyridin-4-ylpiperidin-4-one
Formula:
C₁₀H₁₂N₂O MW: 176,2151 g/mol Storage Temperature: Ambient |
MDL Number:
MFCD09030756 CAS Number: 126832-81-3 |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...