3,5-Dibromo-2,6-dimethylpyridine 98%

Supplier: Apollo Scientific
Warning

OR907091-1G OR907091-5G
APOSOR907091-1GEA 188 EUR
APOSOR907091-1G APOSOR907091-5G
3,5-Dibromo-2,6-dimethylpyridine 98%
3,5-Dibromo-2,6-dimethylpyridine
Formula: C7H7Br2N
MW: 264,950 g/mol
Melting Pt: 65 °C
Storage Temperature: Refrigerator
MDL Number: MFCD00234282
CAS Number: 3430-34-0

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR