1-Methyl-1H-pyrrolo[2,3-b]pyridine 95%

Supplier: Apollo Scientific

OR907187-1G OR907187-5G
APOSOR907187-1GEA 119 EUR
APOSOR907187-1G APOSOR907187-5G
1-Methyl-1H-pyrrolo[2,3-b]pyridine 95%
1-Methyl-1H-pyrrolo[2,3-b]pyridine
Formula: C₈H₈N₂
MW: 132,1600 g/mol
Storage Temperature: Ambient
MDL Number: MFCD09743451
CAS Number: 27257-15-4

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR