4-Amino-3-chloro-5-methylpyridine 96%

Supplier: Apollo Scientific

Synonyms: 3-Chloro-5-methylpyridin-4-amine

OR907324-1G OR907324-250MG
APOSOR907324-1GEA 481 EUR
APOSOR907324-1G APOSOR907324-250MG
4-Amino-3-chloro-5-methylpyridine 96%
4-Amino-3-chloro-5-methylpyridine
Formula: C₆H₇ClN₂
MW: 142,5862 g/mol
Storage Temperature: Ambient
MDL Number: MFCD08277270
CAS Number: 97944-42-8

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR