5,6,7,8-Tetrahydro-2-naphthaleneboronic acid 98%
Supplier: Apollo Scientific
Synonyms:
(5,6,7,8-Tetrahydronaphthalen-2-yl)boronic acid
OR907340-1G
OR907340-250MG
APOSOR907340-1GEA
539
EUR
APOSOR907340-1G
APOSOR907340-250MG
5,6,7,8-Tetrahydro-2-naphthaleneboronic acid 98%
5,6,7,8-Tetrahydro-2-naphthaleneboronic acid
Formula:
C₁₀H₁₃BO₂ MW: 176,0200 g/mol Storage Temperature: Ambient |
MDL Number:
MFCD11183308 CAS Number: 405888-56-4 |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...