Methyl-4-hydroxypicolinate 98%

Supplier: Apollo Scientific

Synonyms: Methyl 4-hydroxypicolinate

OR907529-1G OR907529-250MG
APOSOR907529-1GEA 364 EUR
APOSOR907529-1G APOSOR907529-250MG
Methyl-4-hydroxypicolinate 98%
Methyl 4-hydroxypicolinate
Formula: C₇H₇NO₃
MW: 153,14 g/mol
Storage Temperature: Ambient
MDL Number: MFCD12025998
CAS Number: 473269-77-1

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR