Methyl-3-methoxy-5-methylbenzoate 96%

Supplier: Apollo Scientific

Synonyms: Methyl 3-methoxy-5-methylbenzoate

OR907669-1G OR907669-250MG
APOSOR907669-1GEA 539 EUR
APOSOR907669-1G APOSOR907669-250MG
Methyl-3-methoxy-5-methylbenzoate 96%
Methyl 3-methoxy-5-methylbenzoate
Formula: C₁₀H₁₂O₃
MW: 180,2000 g/mol
Storage Temperature: Ambient
MDL Number: MFCD09833214
CAS Number: 108593-44-8

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR