Methyl-3,5-dimethylbenzoate 97%
Supplier: Apollo Scientific
Synonyms:
Methyl 3,5-dimethylbenzoate
OR907873-100G
APOSOR907873-100GEA
134
EUR
APOSOR907873-100G
Methyl-3,5-dimethylbenzoate 97%
Methyl 3,5-dimethylbenzoate
Formula:
C₁₀H₁₂O₂ MW: 164,21 g/mol Storage Temperature: Ambient |
MDL Number:
MFCD00077971 CAS Number: 25081-39-4 |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...