2-Amino-3-methylbenzamide 98%

Supplier: Apollo Scientific
Warning

OR908166-1G OR908166-25G OR908166-5G
APOSOR908166-1GEA 74.1 EUR
APOSOR908166-1G APOSOR908166-25G APOSOR908166-5G
2-Amino-3-methylbenzamide 98%
2-Amino-3-methylbenzamide
Formula: C₈H₁₀N₂O
MW: 150,1800 g/mol
Storage Temperature: Ambient
MDL Number: MFCD04034517
CAS Number: 1885-32-1

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR