4-tert-Butylbenzylamine 98%
Supplier: Apollo Scientific
|
Danger
|
Synonyms:
p-tert-Butylbenzylamine
OR9087-1G
OR9087-25G
OR9087-5G
OR9087-100G
APOSOR9087-1GEA
27.7
EUR
APOSOR9087-1G
APOSOR9087-25G
APOSOR9087-5G
APOSOR9087-100G
4-tert-Butylbenzylamine 98%
4-tert-Butylbenzylamine
Formula:
C₁₁H₁₇N MW: 163,26 g/mol Boiling Pt: 124 °C Density: 0,927 g/cm³ (25 °C) Flash Pt: 107 °C Storage Temperature: Freezer |
MDL Number:
MFCD00040754 CAS Number: 39895-55-1 UN: 2735 ADR: 8,III |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...