5-(4,4,5,5-Tetramethyl-1,3,2-dioxaborolan-2-yl)isothiazole ≥98%

Supplier: Apollo Scientific
Warning

OR908739-100MG OR908739-250MG
APOSOR908739-100MGEA 231 EUR
APOSOR908739-100MG APOSOR908739-250MG
5-(4,4,5,5-Tetramethyl-1,3,2-dioxaborolan-2-yl)isothiazole ≥98%
5-(4,4,5,5-Tetramethyl-1,3,2-dioxaborolan-2-yl)isothiazole
Formula: C₉H₁₄BNO₂S
Storage Temperature: Ambient
MDL Number: MFCD08061126
CAS Number: 1045809-78-6

Order Now

SPECIFICATIONS

Environmentally Preferable

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR