2,4,4,5,5-Pentamethyl-1,3,2-dioxaborolane 98%

Supplier: Apollo Scientific

Synonyms: Methylboronic acid pinacol ester

OR908873-25G
APOSOR908873-25GEA 87 EUR
APOSOR908873-25G
2,4,4,5,5-Pentamethyl-1,3,2-dioxaborolane 98%
2,4,4,5,5-Pentamethyl-1,3,2-dioxaborolane
Formula: C₇H₁₅BO₂
MW: 142,01 g/mol
Storage Temperature: Ambient
MDL Number: MFCD09027082
CAS Number: 94242-85-0

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR