5-Amino-6-chloroquinoline 98%

Supplier: Apollo Scientific

OR909002-1G OR909002-5G
APOSOR909002-1GEA 40.6 EUR
APOSOR909002-1G APOSOR909002-5G
5-Amino-6-chloroquinoline 98%
5-Amino-6-chloroquinoline
Formula: C₉H₇ClN₂
MW: 178.6200 g/mol
Storage Temperature: Ambient
MDL Number: MFCD02947117
CAS Number: 341010-40-0

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR