trans-3,4,4-Trimethyl-2-pentenoic acid 98%
Supplier: Apollo Scientific
OR909227-1G
OR909227-5G
APOSOR909227-1GEA
168
EUR
APOSOR909227-1G
APOSOR909227-5G
trans-3,4,4-Trimethyl-2-pentenoic acid 98%
trans-3,4,4-Trimethyl-2-pentenoic acid
Formula:
C₈H₁₄O₂ Storage Temperature: Ambient |
MDL Number:
MFCD11172805 CAS Number: 99799-04-9 |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...