2-Chloro-4-piperidin-1-ylpyrimidine 98%
Supplier: Apollo Scientific
OR909497-1G
OR909497-5G
APOSOR909497-1GEA
258
EUR
APOSOR909497-1G
APOSOR909497-5G
2-Chloro-4-piperidin-1-ylpyrimidine 98%
2-Chloro-4-piperidin-1-ylpyrimidine
Formula:
C₉H₁₂ClN₃ MW: 197,67 g/mol Storage Temperature: Ambient |
MDL Number:
MFCD01912571 CAS Number: 5429-00-5 |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...