5-Methoxy-2-tetralone 97%
Supplier: Apollo Scientific
|
Warning
|
Synonyms:
3,4-Dihydro-5-methoxy-2(1H)-naphthalenone
OR9095-1G
OR9095-5G
APOSOR9095-1GEA
14.7
EUR
APOSOR9095-1G
APOSOR9095-5G
5-Methoxy-2-tetralone 97%
5-Methoxy-2-tetralone
Formula:
C₁₁H₁₂O₂ MW: 176,22 g/mol Boiling Pt: 165 °C/10mmHg Melting Pt: 32…36 °C Flash Pt: >110 °C Storage Temperature: Ambient |
MDL Number:
MFCD00064958 CAS Number: 32940-15-1 |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...