1-(3-Chlorophenyl)piperidine 97%

Supplier: Apollo Scientific

OR909839-10G OR909839-1G OR909839-5G
APOSOR909839-10GEA 720 EUR
APOSOR909839-10G APOSOR909839-1G APOSOR909839-5G
1-(3-Chlorophenyl)piperidine 97%
1-(3-Chlorophenyl)piperidine
Formula: C₁₁H₁₄ClN
Storage Temperature: Ambient
MDL Number: MFCD10699658
CAS Number: 102563-84-8

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR