5-(3-Chlorophenyl)isoxazole 98%

Supplier: Apollo Scientific

OR909965-1G OR909965-5G
APOSOR909965-1GEA 174 EUR
APOSOR909965-1G APOSOR909965-5G
5-(3-Chlorophenyl)isoxazole 98%
5-(3-Chlorophenyl)isoxazole
Formula: C₉H₆ClNO
Storage Temperature: Ambient
MDL Number: MFCD03232871
CAS Number: 7064-34-8

Order Now

SPECIFICATIONS

Environmentally Preferable

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR