3-(4-Chlorophenyl)isoxazole 95%
Supplier: Apollo Scientific
OR909998-10G
OR909998-1G
OR909998-5G
APOSOR909998-10GEA
754
EUR
APOSOR909998-10G
APOSOR909998-1G
APOSOR909998-5G
3-(4-Chlorophenyl)isoxazole 95%
3-(4-Chlorophenyl)isoxazole
Formula:
C₉H₆ClNO Storage Temperature: Ambient |
MDL Number:
MFCD08446383 CAS Number: 31301-39-0 |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...