5-Bromo-2-chlorothiazole 97%
Supplier: Apollo Scientific
OR910270-1G
OR910270-250MG
OR910270-5G
APOSOR910270-1GEA
105
EUR
APOSOR910270-1G
APOSOR910270-250MG
APOSOR910270-5G
5-Bromo-2-chlorothiazole 97%
5-Bromo-2-chlorothiazole
Formula:
C₃HBrClNS MW: 198,47 g/mol Storage Temperature: Ambient |
MDL Number:
MFCD14702782 CAS Number: 3034-56-8 |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...