5-Phenyl-1,3,4-oxadiazol-2-ol 98%

Supplier: Apollo Scientific

Synonyms: 5-Phenyl-1,3,4-oxadiazol-2(3H)-one

OR910413-1G
APOSOR910413-1GEA 142 EUR
APOSOR910413-1G
5-Phenyl-1,3,4-oxadiazol-2-ol 98%
5-Phenyl-1,3,4-oxadiazol-2-ol
Formula: C₈H₆N₂O₂
MW: 162,15 g/mol
Storage Temperature: Ambient
MDL Number: MFCD05023177
CAS Number: 1199-02-6

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR