6-Carboxyoxindole 97%

Supplier: Apollo Scientific

OR910478-1G OR910478-5G
APOSOR910478-1GEA 85.8 EUR
APOSOR910478-1G APOSOR910478-5G
6-Carboxyoxindole 97%
6-Carboxyoxindole
Formula: C₉H₇NO₃
Storage Temperature: Ambient
MDL Number: MFCD01415801
CAS Number: 334952-09-9

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR