5-Hydroxyoxindole 95%

Supplier: Apollo Scientific

Synonyms: 5-Hydroxyindolin-2-one

OR910617-1G OR910617-5G
APOSOR910617-1GEA 78.3 EUR
APOSOR910617-1G APOSOR910617-5G
5-Hydroxyoxindole 95%
5-Hydroxyoxindole
Formula: C₈H₇NO₂
MW: 149.15 g/mol
Storage Temperature: Ambient
MDL Number: MFCD00014566
CAS Number: 3416-18-0

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR