2-Phenylbenzonitrile 98%

Supplier: Apollo Scientific

Synonyms: [1,1'-Biphenyl]-2-carbonitrile

OR910733-1G OR910733-5G
APOSOR910733-1GEA 139 EUR
APOSOR910733-1G APOSOR910733-5G
2-Phenylbenzonitrile 98%
2-Phenylbenzonitrile
Formula: C₁₃H₉N
MW: 179,2173 g/mol
Storage Temperature: Ambient
MDL Number: MFCD00060669
CAS Number: 24973-49-7

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR