Methylmalonic acid 97%

Supplier: Apollo Scientific

Synonyms: 2-Methylmalonic acid

OR911347-100G OR911347-25G
APOSOR911347-100GEA 111 EUR
APOSOR911347-100G APOSOR911347-25G
Methylmalonic acid 97%
Methylmalonic acid
Formula: C₄H₆O₄
MW: 118,09 g/mol
Storage Temperature: Ambient
MDL Number: MFCD00002656
CAS Number: 516-05-2

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR