Dimethylmalonic acid 98%

Supplier: Apollo Scientific

Synonyms: 2,2-Dimethylmalonic acid

OR911410-100G OR911410-25G OR911410-500G
APOSOR911410-100GEA 109 EUR
APOSOR911410-100G APOSOR911410-25G APOSOR911410-500G
Dimethylmalonic acid 98%
Dimethylmalonic acid
Formula: C₅H₈O₄
MW: 132,12 g/mol
Storage Temperature: Ambient
MDL Number: MFCD00004193
CAS Number: 595-46-0

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR