2,4,6-Trimethylbenzaldehyde 95%

Supplier: Apollo Scientific

Synonyms: Mesitaldehyde

OR911451-100G OR911451-500G
APOSOR911451-100GEA 116 EUR
APOSOR911451-100G APOSOR911451-500G
2,4,6-Trimethylbenzaldehyde 95%
2,4,6-Trimethylbenzaldehyde
Formula: C₁₀H₁₂O
MW: 148.2 g/mol
Boiling Pt: 237 °C/760mmHg
Melting Pt: 11 °C
Density: 1.02 g/cm³ (25 °C)
Flash Pt: 104 °C
Storage Temperature: Ambient
MDL Number: MFCD00003341
CAS Number: 487-68-3

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR