2-(Methylthio)pyrazine 98%
Supplier: Apollo Scientific
Synonyms:
2-Methylthiopyrazine
OR911470-100G
OR911470-25G
APOSOR911470-100GEA
97.4
EUR
APOSOR911470-100G
APOSOR911470-25G
2-(Methylthio)pyrazine 98%
2-(Methylthio)pyrazine
Formula:
C₅H₆N₂S MW: 126,18 g/mol Storage Temperature: Ambient |
MDL Number:
MFCD00041024 CAS Number: 21948-70-9 |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...